siRNA Details
| Gene Symbol: CTSG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001911.2 | NP_001902.1 | 4 | 403 | ||
| XM_011536499.1 | XP_011534801.1 | 4 | 423 | |||
| GenBank mRNA | AK225914.1 | - | 3 | 850 | ||
| BC014460.1 | - | 4 | 403 | 4859911 | ||
| CR456807.1 | - | 4 | 366 | |||
| CR541704.1 | - | 4 | 366 | |||
| M16117.1 | - | 4 | 374 | |||

CTSG
Genomic Map