siRNA Details
| Gene Symbol: KCNJ10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002241.4 | NP_002232.2 | 2 | 1966 | ||
| GenBank mRNA | BC034036.1 | - | 2 | 1868 | ||
| BC131627.1 | - | 2 | 1862 | 40118533 | ||
| U52155.1 | - | 2 | 1952 | |||
| U73193.1 | - | 1 | 1301 | |||

KCNJ10
Genomic Map