siRNA Details
| Gene Symbol: KCNJ13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172416.1 | NP_001165887.1 | 3 | 1753 | ||
| NM_001172417.1 | NP_001165888.1 | 3 | 1756 | |||
| NM_002242.4 | NP_002233.2 | 3 | 1989 | |||
| GenBank mRNA | AJ007557.1 | - | 3 | 1968 | ||
| BC037290.2 | - | 3 | 1989 | 4817527 | ||

KCNJ13
Genomic Map