siRNA Details
| Gene Symbol: MMP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002428.3 | NP_002419.1 | 3, 4 | 1227 | ||
| GenBank mRNA | BC006412.1 | - | 3, 4 | 347 | ||
| BC036495.1 | - | 3, 4 | 1233 | 5263145 | ||
| BC055428.1 | - | 3, 4 | 1228 | 4811572 | ||
| BT006715.1 | - | 1, 2 | 127 | |||
| D86331.1 | - | 2, 3 | 165 | |||
| Z48482.1 | - | 3, 4 | 490 | |||

MMP15
Genomic Map