siRNA Details
Gene Symbol: NDUFA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002490.3 | NP_002481.2 | 3 | 753 | ||
GenBank mRNA | BC002772.1 | - | 3 | 614 | 3619356 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NDUFA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002490.3 | NP_002481.2 | 3 | 753 | ||
GenBank mRNA | BC002772.1 | - | 3 | 614 | 3619356 |