siRNA Details
| Gene Symbol: NDUFB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199957.1 | NP_001186886.1 | 2, 3 | 223 | ||
| NM_002492.3 | NP_002483.1 | 4, 5 | 379 | |||
| GenBank mRNA | AF047181.1 | - | 4, 5 | 356 | ||
| AK313822.1 | - | 4, 5 | 357 | |||
| BC005271.1 | - | 4, 5 | 370 | 3997377 | ||
| BC009796.1 | - | 4, 5 | 374 | 4284776 | ||
| BC017909.1 | - | 4, 5 | 369 | |||
| BC093071.1 | - | 2, 3 | 200 | 30325282 | ||
| BG776403.1 | - | 4, 5 | 386 | |||
| BX648034.1 | - | 2, 3 | 217 | |||
| DA112751.1 | - | 4, 5 | 382 | |||

NDUFB5
Genomic Map