siRNA Details
| Gene Symbol: PGD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304451.1 | NP_001291380.1 | 6 | 668 | ||
| NM_001304452.1 | NP_001291381.1 | 7 | 813 | |||
| NM_002631.3 | NP_002622.2 | 7 | 734 | |||
| GenBank mRNA | AK290404.1 | - | 7 | 659 | ||
| AK296889.1 | - | 8 | 562 | |||
| AK298830.1 | - | 7 | 813 | |||
| AK300952.1 | - | 6 | 571 | |||
| AK304423.1 | - | 6 | 591 | |||
| BC000368.2 | - | 7 | 645 | 2819330 | ||
| U30255.1 | - | 6 | 614 | |||

PGD
Genomic Map