siRNA Details
| Gene Symbol: PRSS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002773.4 | NP_002764.1 | 6 | 1588 | ||
| GenBank mRNA | AK296656.1 | - | 6 | 1300 | ||
| BC001462.2 | - | 6 | 1499 | 3138532 | ||
| L41351.1 | - | 6 | 1552 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRSS8| Gene Symbol: PRSS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002773.4 | NP_002764.1 | 6 | 1588 | ||
| GenBank mRNA | AK296656.1 | - | 6 | 1300 | ||
| BC001462.2 | - | 6 | 1499 | 3138532 | ||
| L41351.1 | - | 6 | 1552 | |||