siRNA Details
| Gene Symbol: PSMB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002799.3 | NP_002790.1 | 1007 | |||
| GenBank mRNA | AJ420455.1 | - | 685 | |||
| BC000509.2 | - | 963 | 2822305 | |||
| GU727627.1 | - | 976 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PSMB7| Gene Symbol: PSMB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002799.3 | NP_002790.1 | 1007 | |||
| GenBank mRNA | AJ420455.1 | - | 685 | |||
| BC000509.2 | - | 963 | 2822305 | |||
| GU727627.1 | - | 976 | ||||