siRNA Details
Gene Symbol: SDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | D10245.1 | - | 1 | 120 | ||
M32246.1 | - | 1 | 120 | |||
U17248.1 | - | 2 | 308 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | D10245.1 | - | 1 | 120 | ||
M32246.1 | - | 1 | 120 | |||
U17248.1 | - | 2 | 308 |