siRNA Details
| Gene Symbol: KIF3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002254.6 | NP_002245.4 | 8 | 4619 | ||
| XM_005264299.2 | XP_005264356.2 | 8 | 4616 | |||
| GenBank mRNA | AF035621.1 | - | 8 | 4116 | ||
| BC130423.1 | - | 8 | 4190 | 40146454 | ||
| BC132783.1 | - | 8 | 4190 | 40146805 | ||
| BC144223.1 | - | 8 | 4184 | 9052743 | ||
| BX571741.1 | - | 8 | 4540 | |||

KIF3C
Genomic Map