siRNA Details
| Gene Symbol: UQCRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003365.2 | NP_003356.2 | 9 | 1136 | ||
| GenBank mRNA | AK129903.1 | - | 9 | 1116 | ||
| AK300694.1 | - | 9 | 1071 | |||
| AK313090.1 | - | 9 | 1106 | |||
| BC009586.1 | - | 9 | 1107 | 4096628 | ||
| D26485.1 | - | 9 | 1098 | |||
| L16842.1 | - | 9 | 1508 | |||

UQCRC1
Genomic Map