siRNA Details
| Gene Symbol: VDAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003374.2 | NP_003365.1 | 9 | 1716 | ||
| NR_036624.1 | 9 | 1794 | ||||
| NR_036625.1 | 9 | 1797 | ||||
| XM_005272075.3 | XP_005272132.1 | 9 | 1713 | |||
| XM_017009821.1 | XP_016865310.1 | 9 | 1644 | |||
| XM_017009822.1 | XP_016865311.1 | 9 | 1623 | |||
| XM_017009823.1 | XP_016865312.1 | 9 | 1646 | |||
| GenBank mRNA | AK095989.1 | - | 9 | 1716 | ||
| AK122953.1 | - | 12 | 3119 | |||
| BC008482.1 | - | 9 | 1501 | 4332199 | ||
| BC071168.1 | - | 9 | 1533 | 6023095 | ||
| BC090042.1 | - | 9 | 1527 | 30395350 | ||
| L06132.1 | - | 9 | 1570 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_011694, AK153094, AK169282, AK207573, AK146239, and others.

VDAC1
Genomic Map