siRNA Details
| Gene Symbol: PRSS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003619.3 | NP_003610.2 | 1, 2 | 556 | ||
| XM_005263318.4 | XP_005263375.1 | 1, 2 | 829 | |||
| XM_011532387.2 | XP_011530689.1 | 1, 2 | 831 | |||
| GenBank mRNA | AJ001531.1 | - | 1, 2 | 549 | ||
| BC007761.2 | - | 1, 2 | 556 | 4126901 | ||
| BG386264.1 | - | 1, 2 | 410 | |||

PRSS12
Genomic Map