siRNA Details
| Gene Symbol: NDUFB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168331.1 | NP_001161803.1 | 2 | 1415 | ||
| NM_004547.5 | NP_004538.2 | 3 | 465 | |||
| GenBank mRNA | AF044957.1 | - | 3 | 394 | ||
| BC000855.1 | - | 3 | 437 | 3459215 | ||
| BC070285.1 | - | 3 | 404 | 6714676 | ||
| BC146885.1 | - | 3 | 436 | 9056813 | ||
| BC146887.1 | - | 3 | 436 | 9056815 | ||
| BF978138.1 | - | 463 | ||||
| CB215975.1 | - | 1 | 373 | |||

NDUFB4
Genomic Map