siRNA Details
| Gene Symbol: KCNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004770.2 | NP_004761.2 | 3 | 3004 | ||
| XM_017013981.1 | XP_016869470.1 | 2 | 2982 | |||
| XM_017013982.1 | XP_016869471.1 | 2 | 2114 | |||
| GenBank mRNA | AF338730.1 | - | 3 | 3004 | ||
| AF450111.1 | - | 2 | 2416 | |||
| AK311263.1 | - | 2 | 667 | |||

KCNB2
Genomic Map