siRNA Details
| Gene Symbol: TP53I3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004881.4 | NP_004872.2 | 4 | 1656 | ||
| NM_147184.3 | NP_671713.1 | 5 | 1257 | |||
| XM_005264650.2 | XP_005264707.1 | 5 | 1129 | |||
| GenBank mRNA | AF010309.1 | - | 4 | 1294 | ||
| AK223382.1 | - | 4 | 1493 | |||
| AK297579.1 | - | 4 | 1097 | |||
| BC000474.2 | - | 5 | 827 | 2961571 | ||
| BC018819.2 | - | 5 | 827 | |||
| BT007149.1 | - | 4 | 767 | |||
| CN285566.1 | - | 1 | 98 | |||

TP53I3
Genomic Map