siRNA Details
| Gene Symbol: NDUFA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005002.4 | NP_004993.1 | 7, 8 | 752 | ||
| GenBank mRNA | AF050641.1 | - | 7, 8 | 742 | ||
| AK291067.1 | - | 7, 8 | 729 | |||
| BC003351.1 | - | 6, 7 | 606 | |||
| BC009311.2 | - | 7, 8 | 733 | 4106931 | ||
| BC015837.1 | - | 7, 8 | 748 | 4293690 | ||
| BC111546.1 | - | 7, 8 | 724 | 5761015 | ||
| L04490.1 | - | 7, 8 | 718 | |||

NDUFA9
Genomic Map