siRNA Details
| Gene Symbol: CAPN15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005632.2 | NP_005623.1 | 14 | 4191 | ||
| XM_011522620.2 | XP_011520922.1 | 15 | 7514 | |||
| XM_011522621.2 | XP_011520923.1 | 15 | 8442 | |||
| XM_011522622.1 | XP_011520924.1 | 13 | 7146 | |||
| XM_011522623.2 | XP_011520925.1 | 16 | 5217 | |||
| XM_011522624.2 | XP_011520926.1 | 15 | 4555 | |||
| XM_011522625.2 | XP_011520927.1 | 14 | 4441 | |||
| XM_011522626.2 | XP_011520928.1 | 15 | 7982 | |||
| XM_011522627.2 | XP_011520929.1 | 15 | 7983 | |||
| XM_011522628.2 | XP_011520930.1 | 15 | 5075 | |||
| XM_011522629.2 | XP_011520931.1 | 15 | 4938 | |||
| XM_011522630.2 | XP_011520932.1 | 14 | 4276 | |||
| XM_011522631.2 | XP_011520933.1 | 13 | 4162 | |||
| XM_011522632.2 | XP_011520934.1 | 13 | 4593 | |||
| XM_017023596.1 | XP_016879085.1 | 16 | 5354 | |||
| GenBank mRNA | BC001869.1 | - | 2 | 917 | ||
| BC021854.1 | - | 10 | 2258 | |||
| BC040169.1 | - | 1 | 387 | |||
| BC112918.1 | - | 6 | 2416 | |||
| U69557.1 | - | 2 | 877 | |||

CAPN15
Genomic Map