siRNA Details
| Gene Symbol: HS3ST3B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006041.2 | NP_006032.1 | 2 | 1678 | ||
| NR_130138.1 | 2 | 1678 | ||||
| XM_017025479.1 | XP_016880968.1 | 2 | 2106 | |||
| GenBank mRNA | AF105377.1 | - | 2 | 1539 | ||
| AK023723.1 | - | 2 | 1645 | |||
| BC063301.1 | - | 2 | 1595 | 30343352 | ||
| BC069664.1 | - | 2 | 1479 | 7262483 | ||
| BC069725.1 | - | 2 | 1478 | 7262495 | ||

HS3ST3B1
Genomic Map