siRNA Details
| Gene Symbol: GZMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006144.3 | NP_006135.1 | 2 | 166 | ||
| GenBank mRNA | BC015739.1 | - | 2 | 137 | 4849202 | |
| BP330335.1 | - | 2 | 166 | |||
| CR456968.1 | - | 2 | 129 | |||
| M18737.1 | - | 2 | 129 | |||

GZMA
Genomic Map