siRNA Details
| Gene Symbol: NPEPPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006310.3 | NP_006301.3 | 13 | 1705 | ||
| XM_011525496.1 | XP_011523798.1 | 14 | 1507 | |||
| XM_017025372.1 | XP_016880861.1 | 13 | 1667 | |||
| XM_017025373.1 | XP_016880862.1 | 14 | 1507 | |||
| GenBank mRNA | AJ132583.1 | - | 13 | 1435 | ||
| AK095877.1 | - | 14 | 1471 | |||
| AK096709.1 | - | 15 | 1492 | |||
| AK293450.1 | - | 7 | 637 | |||
| AK293995.1 | - | 6 | 619 | |||
| AK296887.1 | - | 12 | 1348 | |||
| AK297107.1 | - | 12 | 1209 | |||
| AK303037.1 | - | 14 | 1500 | |||
| BC065294.1 | - | 13 | 1394 | 6059589 | ||

NPEPPS
Genomic Map