siRNA Details
| Gene Symbol: VAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006373.3 | NP_006364.2 | 6 | 1360 | ||
| GenBank mRNA | AK097072.1 | - | 7 | 1187 | ||
| AK290460.1 | - | 6 | 1336 | |||
| AK298536.1 | - | 6 | 1070 | |||
| AK315838.1 | - | 6 | 924 | |||
| BC001913.1 | - | 6 | 1229 | |||
| BC008725.2 | - | 6 | 1284 | 3162543 | ||
| BC014279.2 | - | 6 | 1266 | 3834206 | ||
| BC015041.2 | - | 6 | 1298 | 3919901 | ||
| BC090858.1 | - | 6 | 1298 | 6528247 | ||
| U18009.1 | - | 6 | 998 | |||
| U25779.1 | - | 6 | 968 | |||

VAT1
Genomic Map