siRNA Details
| Gene Symbol: USP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008563.4 | NP_001008563.2 | 10 | 980 | ||
| NM_001110303.3 | NP_001103773.2 | 10 | 882 | |||
| NM_006676.7 | NP_006667.3 | 10 | 882 | |||
| XM_005251665.3 | XP_005251722.1 | 10 | 870 | |||
| XM_011518161.2 | XP_011516463.1 | 11 | 1222 | |||
| XM_011518162.2 | XP_011516464.1 | 10 | 1401 | |||
| GenBank mRNA | AB023220.1 | - | 9 | 803 | ||
| AY074877.1 | - | 10 | 919 | |||
| BC039593.1 | - | 10 | 829 | 5588842 | ||
| BX462233.2 | - | 2 | 144 | |||

USP20
Genomic Map