siRNA Details
| Gene Symbol: CLCA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012128.3 | NP_036260.2 | 5 | 755 | ||
| NR_024602.1 | 4 | 646 | ||||
| XM_011541015.2 | XP_011539317.1 | 5 | 574 | |||
| GenBank mRNA | AF127035.1 | - | 5 | 739 | ||
| AK000072.1 | - | 5 | 753 | |||
| AK307785.1 | - | 4 | 591 | |||
| AK307898.1 | - | 3 | 360 | |||
| AK314430.1 | - | 5 | 751 | |||
| AY358470.1 | - | 5 | 735 | |||
| BC113687.1 | - | 5 | 750 | 8322739 | ||
| BC113689.1 | - | 5 | 749 | 8322741 | ||

CLCA4
Genomic Map