siRNA Details
| Gene Symbol: KCNH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001314030.1 | NP_001300959.1 | 6, 7 | 1236 | ||
| NM_012284.2 | NP_036416.1 | 6, 7 | 1240 | |||
| XM_011538085.2 | XP_011536387.1 | 6, 7 | 1856 | |||
| XM_011538086.2 | XP_011536388.1 | 6, 7 | 1852 | |||
| XM_017019096.1 | XP_016874585.1 | 5, 6 | 1622 | |||
| XM_017019097.1 | XP_016874586.1 | 6, 7 | 860 | |||
| GenBank mRNA | AB022696.1 | - | 6, 7 | 1015 | ||
| AB033108.1 | - | 6, 7 | 1240 | |||
| BC150602.1 | - | 6, 7 | 1121 | 9056972 | ||

KCNH3
Genomic Map