siRNA Details
| Gene Symbol: KCNH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001314030.1 | NP_001300959.1 | 12 | 2420 | ||
| NM_012284.2 | NP_036416.1 | 12 | 2424 | |||
| XM_011538085.2 | XP_011536387.1 | 12 | 3040 | |||
| XM_011538086.2 | XP_011536388.1 | 12 | 3036 | |||
| XM_017019096.1 | XP_016874585.1 | 11 | 2806 | |||
| XM_017019097.1 | XP_016874586.1 | 12 | 2044 | |||
| GenBank mRNA | AB022696.1 | - | 12 | 2199 | ||
| AB033108.1 | - | 12 | 2424 | |||

KCNH3
Genomic Map