siRNA Details
| Gene Symbol: KCNK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017424.2 | NP_001017424.1 | 2 | 558 | ||
| NM_001017425.2 | NP_001017425.2 | 2 | 359 | |||
| NM_014217.3 | NP_055032.1 | 2 | 235 | |||
| XM_011509522.2 | XP_011507824.1 | 2 | 1761 | |||
| XM_011509523.1 | XP_011507825.1 | 2 | 163 | |||
| XM_011509524.2 | XP_011507826.1 | 2 | 210 | |||
| XM_017001248.1 | XP_016856737.1 | 3 | 946 | |||
| XM_017001249.1 | XP_016856738.1 | 3 | 897 | |||
| GenBank mRNA | AF004711.1 | - | 2 | 343 | ||
| AF129399.1 | - | 1 | 145 | |||
| AF171068.1 | - | 2 | 159 | |||
| AK123409.1 | - | 2 | 247 | |||
| AK291483.1 | - | 2 | 339 | |||
| AK294780.1 | - | 2 | 341 | |||
| AK315249.1 | - | 3 | 325 | |||
| AU280191.1 | - | 2 | 301 | |||
| AY552980.2 | - | 2 | 245 | |||
| AY552981.1 | - | 2 | 250 | |||
| BC069462.1 | - | 2 | 159 | 7262344 | ||
| BC101693.1 | - | 2 | 204 | 8069199 | ||
| BC101695.1 | - | 2 | 204 | 8069201 | ||
| BC143586.1 | - | 2 | 205 | 9052097 | ||
| CD625291.1 | - | 2 | 554 | |||
| CD625297.1 | - | 2 | 552 | |||
| CD625301.1 | - | 2 | 173 | |||
| EF165334.1 | - | 2 | 292 | |||
| EF165335.1 | - | 2 | 257 | |||
| KF182338.1 | - | 2 | 194 | |||
| KF182339.1 | - | 2 | 168 | |||

KCNK2
Genomic Map