siRNA Details
| Gene Symbol: CAPN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014296.2 | NP_055111.1 | 18, 19 | 2338 | ||
| XM_011533555.2 | XP_011531857.1 | 19, 20 | 2237 | |||
| XM_011533556.2 | XP_011531858.1 | 20, 21 | 2469 | |||
| XM_011533557.2 | XP_011531859.1 | 17, 18 | 2077 | |||
| XM_017006043.1 | XP_016861532.1 | 19, 20 | 2421 | |||
| XM_017006044.1 | XP_016861533.1 | 16, 17 | 2029 | |||
| XM_017006045.1 | XP_016861534.1 | 17, 18 | 2095 | |||
| GenBank mRNA | AB028639.1 | - | 18, 19 | 2319 | ||
| AK093742.1 | - | 1, 2 | 903 | |||
| AK314253.1 | - | 18, 19 | 2300 | |||
| BC056202.1 | - | 18, 19 | 2316 | 5274889 | ||

CAPN7
Genomic Map