siRNA Details
| Gene Symbol: TRPM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014555.3 | NP_055370.1 | 7 | 958 | ||
| XM_017017628.1 | XP_016873117.1 | 8 | 1045 | |||
| GenBank mRNA | AF177473.1 | - | 7 | 958 | ||
| AJ270996.1 | - | 7 | 931 | |||
| BC093787.1 | - | 7 | 955 | 7939632 | ||
| BC093789.1 | - | 7 | 955 | 7939634 | ||
| BC143351.1 | - | 7 | 955 | 9051856 | ||
| BC143353.1 | - | 7 | 955 | 9051858 | ||

TRPM5
Genomic Map