siRNA Details
| Gene Symbol: USP33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015017.4 | NP_055832.3 | 5, 6 | 657 | ||
| NM_201624.2 | NP_963918.1 | 4, 5 | 478 | |||
| NM_201626.2 | NP_963920.1 | 5, 6 | 657 | |||
| XM_005270648.1 | XP_005270705.1 | 4, 5 | 305 | |||
| XM_005270649.1 | XP_005270706.1 | 4, 5 | 305 | |||
| XM_011541055.1 | XP_011539357.1 | 4, 5 | 305 | |||
| XM_011541056.2 | XP_011539358.1 | 4, 5 | 305 | |||
| XM_017000722.1 | XP_016856211.1 | 4, 5 | 478 | |||
| XM_017000723.1 | XP_016856212.1 | 4, 5 | 429 | |||
| XM_017000724.1 | XP_016856213.1 | 4, 5 | 478 | |||
| XM_017000725.1 | XP_016856214.1 | 4, 5 | 478 | |||
| XM_017000726.1 | XP_016856215.1 | 4, 5 | 478 | |||
| XM_017000727.1 | XP_016856216.1 | 4, 5 | 321 | |||
| XM_017000728.1 | XP_016856217.1 | 4, 5 | 283 | |||
| XM_017000729.1 | XP_016856218.1 | 4, 5 | 305 | |||
| GenBank mRNA | AB029020.1 | - | 4, 5 | 399 | ||
| AF383172.1 | - | 5, 6 | 630 | |||
| AF383173.1 | - | 4, 5 | 451 | |||
| AK022864.1 | - | 5, 6 | 630 | |||
| AK225499.1 | - | 2, 3 | 98 | |||
| AK308402.1 | - | 4, 5 | 369 | |||
| BC016663.1 | - | 5, 6 | 511 | 3874822 | ||
| DB232523.1 | - | 4, 5 | 478 | |||

USP33
Genomic Map