siRNA Details
| Gene Symbol: RDH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015725.2 | NP_056540.2 | 6 | 1194 | ||
| GenBank mRNA | AF229845.1 | - | 6 | 1194 | ||
| AK024022.1 | - | 6 | 959 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RDH8| Gene Symbol: RDH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015725.2 | NP_056540.2 | 6 | 1194 | ||
| GenBank mRNA | AF229845.1 | - | 6 | 1194 | ||
| AK024022.1 | - | 6 | 959 | |||