siRNA Details
| Gene Symbol: DHRS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322280.1 | NP_001309209.1 | 5 | 749 | ||
| NM_001322281.1 | NP_001309210.1 | 5 | 905 | |||
| NM_001322282.1 | NP_001309211.1 | 4 | 658 | |||
| NM_016029.3 | NP_057113.1 | 5 | 898 | |||
| XM_017021368.1 | XP_016876857.1 | 5 | 1006 | |||
| GenBank mRNA | AF126782.1 | - | 844 | |||
| AF151844.1 | - | 5 | 791 | |||
| AK313285.1 | - | 5 | 823 | |||
| AL117420.1 | - | 5 | 834 | |||
| AY359031.1 | - | 5 | 707 | |||
| BC000637.2 | - | 5 | 734 | 3344010 | ||
| BC007337.2 | - | 5 | 752 | 3616802 | ||

DHRS7
Genomic Map