siRNA Details
| Gene Symbol: PRODH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195226.1 | NP_001182155.1 | 6 | 574 | ||
| NM_016335.4 | NP_057419.4 | 7 | 989 | |||
| GenBank mRNA | AB209472.1 | - | 5 | 820 | ||
| AK091063.1 | - | 1 | 1120 | |||
| BC036534.1 | - | 6 | 798 | |||
| BC068260.1 | - | 6 | 790 | |||
| BC094736.1 | - | 6 | 619 | |||
| BC118597.1 | - | 6 | 574 | 40108132 | ||
| BC121809.1 | - | 6 | 574 | 40108133 | ||
| KJ534920.1 | - | 6 | 785 | |||
| U79754.1 | - | 7 | 989 | |||
| Gene Symbol: LOC102724788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006724935.3 | XP_006724998.1 | 6 | 818 | ||
| XM_006724936.3 | XP_006724999.1 | 5 | 728 | |||
| XM_017030166.1 | XP_016885655.1 | 6 | 915 | |||
| XM_017030167.1 | XP_016885656.1 | 5 | 1459 | |||
| XM_017030168.1 | XP_016885657.1 | 6 | 636 | |||
| XM_017030169.1 | XP_016885658.1 | 4 | 1194 | |||
| XM_017030170.1 | XP_016885659.1 | 5 | 545 | |||

PRODH
Genomic Map