siRNA Details
| Gene Symbol: BANK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083907.2 | NP_001077376.2 | 4 | 756 | ||
| NM_001127507.2 | NP_001120979.2 | 3 | 588 | |||
| NM_017935.4 | NP_060405.4 | 4 | 987 | |||
| XM_017008337.1 | XP_016863826.1 | 4 | 734 | |||
| GenBank mRNA | AB063170.1 | - | 3 | 623 | ||
| AK000713.1 | - | 4 | 703 | |||
| AK091523.1 | - | 8 | 1088 | |||
| AK292133.1 | - | 4 | 831 | |||
| AL834291.1 | - | 4 | 806 | |||
| BC032241.1 | - | 4 | 855 | 5214147 | ||
| EU051376.1 | - | 3 | 326 | |||

BANK1
Genomic Map