siRNA Details
| Gene Symbol: CPVL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019029.2 | NP_061902.2 | 4, 5 | 616 | ||
| NM_031311.3 | NP_112601.3 | 4, 5 | 528 | |||
| XM_011515437.1 | XP_011513739.1 | 4, 5 | 695 | |||
| XM_017012366.1 | XP_016867855.1 | 4, 5 | 528 | |||
| GenBank mRNA | AF106704.2 | - | 4, 5 | 448 | ||
| AF217508.1 | - | 3, 4 | 415 | |||
| AF282617.1 | - | 4, 5 | 478 | |||
| AK075433.1 | - | 4, 5 | 529 | |||
| AK124472.1 | - | 8, 9 | 1056 | |||
| AK225545.1 | - | 4, 5 | 479 | |||
| AY358549.1 | - | 4, 5 | 505 | |||
| BC016838.2 | - | 4, 5 | 576 | 3888647 | ||

CPVL
Genomic Map