siRNA Details
| Gene Symbol: CYP26B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277742.1 | NP_001264671.1 | 5 | 3116 | ||
| NM_019885.3 | NP_063938.1 | 6 | 3341 | |||
| XM_005264433.4 | XP_005264490.1 | 6 | 3305 | |||
| XM_011532988.1 | XP_011531290.1 | 5 | 2902 | |||
| GenBank mRNA | AF252297.1 | - | 6 | 3313 | ||
| CR749631.1 | - | 5 | 2965 | |||

CYP26B1
Genomic Map