siRNA Details
Gene Symbol: CACHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293274.1 | NP_001280203.1 | 6 | 1205 | ||
NM_020925.2 | NP_065976.2 | 7 | 926 | |||
XM_011541862.1 | XP_011540164.1 | 6 | 1201 | |||
XM_011541863.1 | XP_011540165.1 | 7 | 1350 | |||
GenBank mRNA | AB046793.1 | - | 5 | 708 | ||
BC039301.1 | - | 7 | 912 | 4836806 | ||
BX537603.1 | - | 6 | 1204 |
