siRNA Details
| Gene Symbol: VAT1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020927.2 | NP_065978.1 | 9 | 2803 | ||
| GenBank mRNA | AB046796.1 | - | 9 | 2773 | ||
| AK001256.1 | - | 1 | 1137 | |||
| AL834240.1 | - | 8 | 2418 | |||
| BC033537.1 | - | 9 | 2781 | 4821265 | ||

VAT1L
Genomic Map