siRNA Details
| Gene Symbol: PRODH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021232.1 | NP_067055.1 | 2 | 265 | ||
| XM_017027084.1 | XP_016882573.1 | 1 | 122 | |||
| XM_017027085.1 | XP_016882574.1 | 1 | 121 | |||
| GenBank mRNA | BC039710.1 | - | 1 | 37 | ||
| U80018.1 | - | 2 | 265 | |||

PRODH2
Genomic Map