siRNA Details
| Gene Symbol: MAGEB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002365.4 | NP_002356.2 | 5 | 997 | ||
| XM_011545513.2 | XP_011543815.1 | 4 | 604 | |||
| XM_011545514.2 | XP_011543816.1 | 3 | 571 | |||
| GenBank mRNA | AK057441.1 | - | 5 | 997 | ||
| BC069784.1 | - | 1 | 319 | 7262601 | ||
| BC074756.2 | - | 1 | 288 | 30915215 | ||
| BC126320.1 | - | 1 | 291 | 8992036 | ||
| BC126322.1 | - | 1 | 291 | 8992038 | ||

MAGEB3
Genomic Map