siRNA Details
| Gene Symbol: DPEP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129758.1 | NP_001123230.1 | 6 | 1060 | ||
| NM_022357.3 | NP_071752.3 | 6 | 1060 | |||
| GenBank mRNA | AJ291679.1 | - | 6 | 974 | ||
| AK057401.1 | - | 6 | 1060 | |||
| AY358390.1 | - | 6 | 983 | |||
| BC037243.1 | - | 6 | 972 | |||
| BC057789.1 | - | 6 | 1001 | 5297958 | ||

DPEP3
Genomic Map