siRNA Details
| Gene Symbol: FAM57A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318006.1 | NP_001304935.1 | 4 | 1013 | ||
| NM_001318007.1 | NP_001304936.1 | 4 | 907 | |||
| NM_001318008.1 | NP_001304937.1 | 3 | 811 | |||
| NM_024792.2 | NP_079068.1 | 5 | 1109 | |||
| GenBank mRNA | AF477201.1 | - | 5 | 1097 | ||
| AK025935.1 | - | 5 | 1048 | |||
| AK125146.1 | - | 4 | 1314 | |||
| BC009729.1 | - | 2 | 602 | |||
| BC026023.1 | - | 4 | 975 | 4548729 | ||
| BX389422.1 | - | 3 | 654 | |||

FAM57A
Genomic Map