siRNA Details
| Gene Symbol: ACAD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136538.1 | NP_001130010.1 | 2 | 338 | ||
| NM_025247.5 | NP_079523.3 | 2 | 338 | |||
| GenBank mRNA | AK092356.1 | - | 2 | 323 | ||
| AK310923.1 | - | 2 | 323 | |||
| AL832043.1 | - | 2 | 316 | |||
| AY323912.1 | - | 2 | 294 | |||
| BC015056.1 | - | 2 | 310 | |||
| BC126358.1 | - | 2 | 252 | 8992074 | ||
| DA146693.1 | - | 2 | 338 | |||

ACAD10
Genomic Map