siRNA Details
| Gene Symbol: KCNK17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135111.1 | NP_001128583.1 | 6 | 1134 | ||
| NM_031460.3 | NP_113648.2 | 5 | 982 | |||
| GenBank mRNA | AF339912.1 | - | 5 | 947 | ||
| AF358910.1 | - | 5 | 842 | |||
| AK315273.1 | - | 5 | 973 | |||
| AY358853.1 | - | 5 | 933 | |||
| BC025726.1 | - | 5 | 929 | 5201326 | ||
| EU978944.1 | - | 5 | 889 | |||

KCNK17
Genomic Map