siRNA Details
| Gene Symbol: PRSS27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318395.1 | NP_001305324.1 | 4 | 856 | ||
| NM_031948.4 | NP_114154.1 | 5 | 1019 | |||
| XM_011522689.2 | XP_011520991.1 | 3 | 748 | |||
| GenBank mRNA | AB056161.1 | - | 5 | 678 | ||
| AJ306593.1 | - | 5 | 647 | |||
| AK055576.1 | - | 2 | 1620 | |||
| AK310214.1 | - | 4 | 531 | |||
| AY030095.1 | - | 5 | 628 | |||
| AY359106.1 | - | 5 | 663 | |||
| BC034294.2 | - | 5 | 683 | 4752547 | ||
| BC036912.2 | - | 4 | 867 | |||

PRSS27
Genomic Map