siRNA Details
| Gene Symbol: USP30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301175.1 | NP_001288104.1 | 8, 9 | 1070 | ||
| NM_032663.4 | NP_116052.2 | 5, 6 | 696 | |||
| XM_005253965.4 | XP_005254022.1 | 4, 5 | 507 | |||
| XM_006719653.3 | XP_006719716.1 | 6, 7 | 769 | |||
| XM_017020048.1 | XP_016875537.1 | 5, 6 | 691 | |||
| XM_017020050.1 | XP_016875539.1 | 5, 6 | 791 | |||
| XM_017020051.1 | XP_016875540.1 | 6, 7 | 647 | |||
| XM_017020052.1 | XP_016875541.1 | 5, 6 | 693 | |||
| XM_017020053.1 | XP_016875542.1 | 4, 5 | 423 | |||
| GenBank mRNA | AJ586136.1 | - | 5, 6 | 587 | ||
| AK027820.1 | - | 5, 6 | 661 | |||
| AK097830.1 | - | 8, 9 | 1070 | |||
| BC004868.2 | - | 5, 6 | 686 | 3833242 | ||
| BC022094.2 | - | 3, 4 | 238 | |||
 Back To Top
 Back To Top
 USP30
USP30 Genomic Map
Genomic Map Back To Top
 Back To Top