siRNA Details
| Gene Symbol: LDHAL6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033195.2 | NP_149972.1 | 1 | 1443 | ||
| GenBank mRNA | AK058192.1 | - | 1 | 1373 | ||
| AY009108.1 | - | 1 | 1416 | |||
| AY642121.1 | - | 1 | 1436 | |||
| BC022034.1 | - | 1 | 1429 | 4825988 | ||
| DB511998.1 | - | 1 | 107 | |||

LDHAL6B
Genomic Map