siRNA Details
| Gene Symbol: CATSPER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053054.3 | NP_444282.3 | 1 | 269 | ||
| GenBank mRNA | AF407333.1 | - | 1 | 131 | ||
| BC032950.2 | - | 1 | 268 | 4822261 | ||
| BC036522.2 | - | 1 | 231 | 5268567 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CATSPER1| Gene Symbol: CATSPER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053054.3 | NP_444282.3 | 1 | 269 | ||
| GenBank mRNA | AF407333.1 | - | 1 | 131 | ||
| BC032950.2 | - | 1 | 268 | 4822261 | ||
| BC036522.2 | - | 1 | 231 | 5268567 | ||