siRNA Details
| Gene Symbol: DPP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139159.4 | NP_631898.3 | 22 | 4178 | ||
| XM_005259673.2 | XP_005259730.1 | 23 | 4287 | |||
| XM_011528405.1 | XP_011526707.1 | 24 | 4339 | |||
| XM_011528406.1 | XP_011526708.1 | 25 | 4449 | |||
| XM_011528408.1 | XP_011526710.1 | 22 | 4124 | |||
| XM_011528410.1 | XP_011526712.1 | 23 | 4339 | |||
| XM_017027453.1 | XP_016882942.1 | 24 | 4929 | |||
| XM_017027454.1 | XP_016882943.1 | 25 | 5039 | |||
| GenBank mRNA | AF452102.2 | - | 22 | 4198 | ||
| AL834376.1 | - | 16 | 3128 | |||
| BC037948.2 | - | 22 | 4115 | 4577856 | ||
| CR627380.1 | - | 14 | 2954 | |||

DPP9
Genomic Map